Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse MafK Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB3809SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3809SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB3809SP Supplier R&D Systems Supplier No. MAB3809SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

MafK Monoclonal specifically detects MafK in Human, Mouse samples. It is validated for Western Blot.

Specifications

Antigen MafK
Applications Western Blot
Classification Monoclonal
Clone 381923
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. O60675
Gene Alias FLJ32205, MGC71717, NFE2U, transcription factor MafK, ubiquitous (p18), v-maf avian musculoaponeurotic fibrosarcoma oncogene family, protein K, v-maf musculoaponeurotic fibrosarcoma (avian) oncogene family, protein K, v-maf musculoaponeurotic fibrosarcoma oncogene homolog K (avian)
Gene Symbols MAFK
Host Species Mouse
Immunogen E. coli-derived recombinant human MafK Thr2-Ser156 Accession # O60675
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7975
Test Specificity Detects human and mouse MafK in Western blots. In Western blots, no cross-reactivity with recombinant human (rh) MafB or rhMafF is observed. In Western blots, prolonged film exposures may slightly detect rhMafG.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.