Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse MCPIP1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB7875SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB7875SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB7875SP Supplier R&D Systems Supplier No. MAB7875SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 5 publications

MCPIP1/ZC3H12A Monoclonal specifically detects MCPIP1/ZC3H12A in Human, Mouse samples. It is validated for Western Blot.

Specifications

Antigen MCPIP1/ZC3H12A
Applications Western Blot
Classification Monoclonal
Clone 604421
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q5D1E8
Gene Alias Reg1, Regnase-1, ZC3H12A, zinc finger CCCH-type containing 12A
Gene Symbols ZC3H12A
Host Species Mouse
Immunogen E. coli-derived recombinant human MCPIP1 Asp426-Glu599 Accession # Q5D1E8
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80149
Test Specificity Detects human MCPIP1 in direct ELISAs and human and mouse MCPIP1 in Western blots. In direct ELISAs, no cross-reactivity with recombinant human MCPIP3 is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human, Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.