Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse MCPIP1 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 5 publications
Supplier: R&D Systems MAB7875
This item is not returnable.
View return policy
Description
MCPIP1/ZC3H12A Monoclonal specifically detects MCPIP1/ZC3H12A in Human, Mouse samples. It is validated for Western Blot.Specifications
MCPIP1/ZC3H12A | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL | |
Q5D1E8 | |
ZC3H12A | |
E. coli-derived recombinant human MCPIP1 Asp426-Glu599 Accession # Q5D1E8 | |
100 μg | |
Primary | |
Detects human MCPIP1 in direct ELISAs and human and mouse MCPIP1 in Western blots. In direct ELISAs, no cross-reactivity with recombinant human MCPIP3 is observed. | |
Human, Mouse | |
Purified |
Western Blot | |
604421 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Reg1, Regnase-1, ZC3H12A, zinc finger CCCH-type containing 12A | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
80149 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction