Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse MEF2C Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB6786SP
This item is not returnable.
View return policy
Description
MEF2C Monoclonal specifically detects MEF2C in Human, Mouse samples. It is validated for Immunohistochemistry, Immunocytochemistry.Specifications
MEF2C | |
Monoclonal | |
LYOPH | |
Immunohistochemistry 5-25 ug/mL, Immunocytochemistry 8-25 ug/mL | |
Q06413 | |
MEF2C | |
E. coli-derived recombinant human MEF2C Ala135-Lys239 Accession # Q06413 | |
25 μg | |
Primary | |
Detects human MEF2C in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human SRF is observed. | |
Human, Mouse | |
Purified |
Immunohistochemistry, Immunocytochemistry | |
681824 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
DEL5q14.3, myocyte enhancer factor 2C, myocyte-specific enhancer factor 2C, polypeptide C | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
4208 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction