Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse MEF2C Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF6786
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF6786 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF6786 Supplier R&D Systems Supplier No. AF6786
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

MEF2C Polyclonal specifically detects MEF2C in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen MEF2C
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.2 ug/mL, Immunohistochemistry 1-15 ug/mL
Gene Accession No. Q06413
Gene Alias DEL5q14.3, myocyte enhancer factor 2C, myocyte-specific enhancer factor 2C, polypeptide C
Gene Symbols MEF2C
Host Species Sheep
Immunogen E. coli-derived recombinant human MEF2C Ala135-Lys239 Accession # Q06413
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4208
Test Specificity Detects human MEF2C in direct ELISAs and Western blots.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Human, Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.