Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse Meprin beta Subunit/MEP1B Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB28951
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB28951 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB28951 Supplier R&D Systems Supplier No. MAB28951
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Meprin beta Subunit/MEP1B Monoclonal specifically detects Meprin beta Subunit/MEP1B in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Meprin beta Subunit/MEP1B
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 289731
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q16820
Gene Alias EC 3.4.24.63, endopeptidase-2, MEP1B, meprin A subunit beta, meprin A, beta, Meprin B, N-benzoyl-L-tyrosyl-p-amino-benzoic acid hydrolase beta subunit, N-benzoyl-L-tyrosyl-P-amino-benzoic acid hydrolase subunit beta, PABA peptide hydrolase, PPH beta
Gene Symbols MEP1B
Host Species Rat
Immunogen Human embryonic kidney cell line HEK293-derived recombinant human Meprin beta Subunit/MEP1B Thr23-Ser293 Accession # Q16820
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4225
Test Specificity Detects the pro form of human or mouse Meprin beta Subunit/MEP1B in Western blots. It does not react with the mature form.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.