Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse Metadherin Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF7180SP
This item is not returnable.
View return policy
Description
LYRIC Polyclonal specifically detects LYRIC in Human, Mouse samples. It is validated for Western Blot.Specifications
LYRIC | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
3D3, AEG1, AEG-13D3/LYRIC, AEG1LYRIC/3D3, Astrocyte elevated gene-1 protein, LYRIC, LYRICMetastasis adhesion protein, Lysine-rich CEACAM1 co-isolated protein, metadherin, MTDH, protein LYRIC | |
Sheep | |
Affinity Purified | |
RUO | |
92140 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 0.25 ug/mL | |
Q80WJ7 | |
MTDH | |
E. coli-derived recombinant mouse Metadherin Lys167-Ser297 Accession # Q80WJ7 | |
25 μg | |
Primary | |
Detects human and mouse Metadherin in direct ELISAs and Western blots. | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction