Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse Metadherin Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF7180
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF7180 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF7180 Supplier R&D Systems Supplier No. AF7180
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

LYRIC Polyclonal specifically detects LYRIC in Human, Mouse samples. It is validated for Western Blot.

Specifications

Antigen LYRIC
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.25 ug/mL
Gene Accession No. Q80WJ7
Gene Alias 3D3, AEG1, AEG-13D3/LYRIC, AEG1LYRIC/3D3, Astrocyte elevated gene-1 protein, LYRIC, LYRICMetastasis adhesion protein, Lysine-rich CEACAM1 co-isolated protein, metadherin, MTDH, protein LYRIC
Gene Symbols MTDH
Host Species Sheep
Immunogen E. coli-derived recombinant mouse Metadherin Lys167-Ser297 Accession # Q80WJ7
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 92140
Test Specificity Detects human and mouse Metadherin in direct ELISAs and Western blots.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Human, Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.