Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse MIF Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $514.50
Specifications
Antigen | MIF |
---|---|
Clone | 932612 |
Dilution | Western Blot 2 ug/mL, Immunohistochemistry 8-25 ug/mL |
Applications | Western Blot, Immunohistochemistry |
Classification | Monoclonal |
Description
MIF Monoclonal specifically detects MIF in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
MIF | |
Western Blot 2 ug/mL, Immunohistochemistry 8-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
EC 5.3.2.1, EC 5.3.3.12, GIFmacrophage migration inhibitory factor, GLIF, Glycosylation-inhibiting factor, L-dopachrome isomerase, L-dopachrome tautomerase, macrophage migration inhibitory factor (glycosylation-inhibiting factor), MMIF, Phenylpyruvate tautomerase | |
MIF | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
932612 | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Mouse | |
Human, Mouse | |
P14174 | |
4282 | |
E. coli-derived recombinant human MIF Met1-Phe114 Accession # P14174 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human MIF in direct ELISA and Western Blots. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title