Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse MMP-24/MT5-MMP Alexa Fluor 700-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems IC924N100UG
This item is not returnable.
View return policy
Description
MMP-24/MT5-MMP Monoclonal specifically detects MMP-24/MT5-MMP in Human, Mouse samples. It is validated for Flow Cytometry.Specifications
MMP-24/MT5-MMP | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q9R0S2 | |
MMP24 | |
E. coli-derived recombinant mouse MMP-24/MT5-MMP Arg125-Gln296 Accession # Q9R0S2 | |
100 μg | |
Primary | |
Detects mouse MMP-24/MT5-MMP in direct ELISAs and detects human and mouse MMP-24/MT5-MMP in Western blots. In Western blots, less than 5% cross-reactivity with recombinant mouse (rm) MMP-3, rmMMP-9, and the catalytic domains of recombinant human MMP-14, 15, and 16 is observed. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG2a |
Flow Cytometry | |
143908 | |
Alexa Fluor 700 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
EC 3.4.24, EC 3.4.24.-, EC 3.4.24.80, matrix metallopeptidase 24 (membrane-inserted), matrix metalloproteinase 24 (membrane-inserted), matrix metalloproteinase-24, membrane-type 5 matrix metalloproteinase, Membrane-type matrix metalloproteinase 5, Membrane-type-5 matrix metalloproteinase, MMP24, MMP-24, MT5MMP, MT5-MMP, MT5-MMPMMP25, MT-MMP 5, MTMMP5, MT-MMP5 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
10893 | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction