Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse MOG Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAM2439
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAM2439 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAM2439 Supplier R&D Systems Supplier No. BAM2439
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

MOG Monoclonal specifically detects MOG in Human, Mouse samples. It is validated for Immunohistochemistry.

Specifications

Antigen MOG
Applications Immunohistochemistry
Classification Monoclonal
Clone 296829
Concentration LYOPH
Conjugate Biotin
Dilution Immunohistochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein. with No Preservative
Gene Accession No. Q61885
Gene Alias MGC26137, MOG Ig-AluB, MOGIG2, myelin oligodendrocyte glycoprotein, myelin-oligodendrocyte glycoprotein
Gene Symbols MOG
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse MOG Gly29-Gly153 Accession # Q61885
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4340
Test Specificity Detects mouse MOG in direct ELISAs. In direct ELISAs, 100% cross-reactivity with recombinant human MOG is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.