Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse MSX1 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 3 publications
Supplier: R&D Systems AF5045
This item is not returnable.
View return policy
Description
MSX1 Polyclonal specifically detects MSX1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
MSX1 | |
Polyclonal | |
Unconjugated | |
P28360 | |
MSX1 | |
E. coli-derived recombinant human MSX1 Met1-Thr165 Accession # P28360 | |
100 μg | |
Primary | |
Detects human and mouse MSX1 in Western blots. | |
Human, Mouse | |
IgG |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
Homeobox protein Hox-7, homeobox protein MSX-1, HOX7, HOX7homeobox 7, HYD1, HYD1msh homeobox homolog 1 (Drosophila), msh (Drosophila) homeo box homolog 1 (formerly homeo box 7), msh homeo box 1, msh homeobox 1, Msh homeobox 1-like protein, msh homeobox homolog 1, OFC5, STHAG1 | |
Goat | |
Affinity Purified | |
RUO | |
4487 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction