Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse Myeloperoxidase/MPO Antibody, R&D Systems™
GSA_VAGREENER_CHOICE

Catalog No. AF3667
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF3667 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF3667 Supplier R&D Systems Supplier No. AF3667
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 37 publications

Myeloperoxidase/MPO Polyclonal specifically detects Myeloperoxidase/MPO in Human, Mouse samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Antigen Myeloperoxidase/MPO
Applications Western Blot, Immunohistochemistry, Immunocytochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.5 ug/mL, Simple Western 5 ug/mL, Immunohistochemistry 1-15 ug/mL, Immunocytochemistry 5-15 ug/mL
Gene Accession No. P11247
Gene Alias EC 1.11.1, EC 1.11.1.7, MPO, myeloperoxidase
Gene Symbols MPO
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse MPO Met16-Thr718 Accession # P11247
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4353
Test Specificity Detects human and mouse Myeloperoxidase/MPO in Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant mouse EPPO is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.