Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse MYF-5 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF4027
This item is not returnable.
View return policy
Description
MYF-5 Polyclonal specifically detects MYF-5 in Human, Mouse samples. It is validated for Immunocytochemistry.Specifications
MYF-5 | |
Polyclonal | |
Unconjugated | |
P13349 | |
MYF5 | |
E. coli-derived recombinant human MYF-5 Met1-Leu255 Accession # P13349 | |
100 μg | |
Primary | |
Detects recombinant human MYF-5 in direct ELISAs and Western blots. | |
Human, Mouse | |
IgG |
Immunocytochemistry | |
LYOPH | |
Immunocytochemistry 5-15 ug/mL | |
BHLHC2, bHLHc2Class C basic helix-loop-helix protein 2, myf-5, MYF5, myogenic factor 5 | |
Goat | |
Affinity Purified | |
RUO | |
4617 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction