Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse Nectin-1 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
$200.50 - $549.50
Specifications
Antigen | Nectin-1/PVRL1 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
Nectin-1/PVRL1 Polyclonal specifically detects Nectin-1/PVRL1 in Human, Mouse samples. It is validated for Western Blot.Specifications
Nectin-1/PVRL1 | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Human, Mouse | |
CD111, CD111 antigen, CLPED1ectodermal dysplasia 4 (Margarita Island type), ED4, Herpes virus entry mediator C, Herpesvirus entry mediator C, Herpesvirus Ig-like receptor, HIgRPVRR, HveC, HVECpoliovirus receptor-like 1, MGC142031, Nectin1, Nectin-1, OFC7nectin 1, poliovirus receptor-related 1 (herpesvirus entry mediator C), poliovirus receptor-related protein 1, PRR, PRR1, PRR1MGC16207, PVRL1, PVRR1, PVRR1nectin-1, SK-12 | |
PVRL1 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
Q15223 | |
5818 | |
Mouse myeloma cell line NS0-derived recombinant human Nectin-1 Gln31-Thr334 Accession # Q15223 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human and mouse Nectin-1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 6% cross-reactivity with recombinant human (rh) Nectin-2, -3, and -4 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title