Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse Neogenin Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 11 publications
Supplier: R&D Systems AF1079SP
This item is not returnable.
View return policy
Description
Neogenin Polyclonal specifically detects Neogenin in Human, Mouse samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Blockade of Receptor-ligand Interaction.Specifications
Neogenin | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
HsT17534, IGDCC2DKFZp547B146, Immunoglobulin superfamily DCC subclass member 2, NEO, NEO1, neogenin, neogenin (chicken) homolog 1, neogenin 1, neogenin homolog 1, NGN, NGNDKFZp547A066 | |
Goat | |
Affinity Purified | |
RUO | |
4756 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunohistochemistry, Blockade of Receptor-ligand Interaction | |
LYOPH | |
Western Blot 0.5 ug/mL, Simple Western 10 ug/mL, Immunohistochemistry 5-15 ug/mL, Blockade of Receptor-ligand Interaction | |
NP_032710 | |
NEO1 | |
Mouse myeloma cell line NS0-derived recombinant mouse Neogenin Ala42-Ile1033 (Asp442-Leu461 del) Accession # NP_032710 | |
25 μg | |
Primary | |
Detects human and mouse Neogenin in direct ELISAs and Western blots. | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction