Learn More
Human/Mouse Neogenin Antibody, R&D Systems™
GREENER_CHOICE
Goat Polyclonal Antibody has been used in 11 publications
Supplier:  R&D Systems AF1079
Catalog No. AF1079
Only null left
Add to Cart
This item is not returnable. View return policy
Description
Description
Neogenin Polyclonal specifically detects Neogenin in Human, Mouse samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Blockade of Receptor-ligand Interaction.
Specifications
Specifications
Neogenin
Polyclonal
Unconjugated
NP_032710
NEO1
Mouse myeloma cell line NS0-derived recombinant mouse Neogenin Ala42-Ile1033 (Asp442-Leu461 del) Accession # NP_032710
100 μg
Primary
Detects human and mouse Neogenin in direct ELISAs and Western blots.
Human, Mouse
IgG
Western Blot, Immunohistochemistry, Blockade of Receptor-ligand Interaction
LYOPH
Western Blot 0.5 ug/mL, Simple Western 10 ug/mL, Immunohistochemistry 5-15 ug/mL, Blockade of Receptor-ligand Interaction
HsT17534, IGDCC2DKFZp547B146, Immunoglobulin superfamily DCC subclass member 2, NEO, NEO1, neogenin, neogenin (chicken) homolog 1, neogenin 1, neogenin homolog 1, NGN, NGNDKFZp547A066
Goat
Affinity Purified
RUO
4756
Reconstitute at 0.2 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.