Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse NeuroD2 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF2775
This item is not returnable.
View return policy
Description
NeuroD2 Polyclonal specifically detects NeuroD2 in Human, Mouse samples. It is validated for Western Blot.Specifications
NeuroD2 | |
Polyclonal | |
Unconjugated | |
Q15784 | |
NEUROD2 | |
E. coli-derived recombinant human NeuroD2 Ser174-Asn382 Accession # Q15784 | |
100 μg | |
Primary | |
Detects human NeuroD2 in direct ELISAs and Western blots. In direct ELISAs, approximately 20% cross-reactivity with recombinant human NeuroD1 is observed. | |
Human, Mouse | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
BHLHA1, bHLHa1neurogenic differentiation factor 2, Class A basic helix-loop-helix protein 1, MGC26304, NDRF, NDRFneurogenic basic-helix-loop-helix protein, NeuroD2, NeuroD-related factor, neurogenic differentiation 2 | |
Goat | |
Affinity Purified | |
RUO | |
4761 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction