Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse Neuronal Pentraxin R/NPTXR Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB4414SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4414SP 25 μg
MAB4414100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB4414SP Supplier R&D Systems Supplier No. MAB4414SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Neuronal Pentraxin R/NPTXR Monoclonal antibody specifically detects Neuronal Pentraxin R/NPTXR in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry

Specifications

Antigen Neuronal Pentraxin R/NPTXR
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 511527
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL, Immunohistochemistry 5-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. O95502
Gene Alias neuronal pentraxin receptor, NPR, NPTXR
Host Species Mouse
Immunogen Mouse myeloma cell line, NS0-derived human Neuronal Pentraxin R/NPTXR, Ala24-Ala500, Accession # O95502
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23467
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.