Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse Neuroplastin 65 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Supplier: R&D Systems AF5360SP
This item is not returnable.
View return policy
Description
Neuroplastin 65 Polyclonal specifically detects Neuroplastin 65 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Neuroplastin 65 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
NPTN | |
Mouse myeloma cell line NS0-derived recombinant human Neuroplastin 65 Gln29-His336 Accession # NP_036560 | |
25 μg | |
Primary | |
Detects human and mouse Neuroplastin 65 in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant human Neuroplastin 55 is observed. | |
Human, Mouse | |
IgG |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
NP_036560 | |
Goat | |
Affinity Purified | |
RUO | |
27020 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction