Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse NFkB1 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF2697SP
This item is not returnable.
View return policy
Description
NFkB1/NFkB p105 Polyclonal specifically detects NFkB1/NFkB p105 in Human, Mouse samples. It is validated for Western Blot, Chromatin Immunoprecipitation (ChIP).Specifications
NFkB1/NFkB p105 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
DNA-binding factor KBF1, EBP-1, NF-kappa-B, NF-kB1, NFKB1, NFKB-p105, Nuclear factor NF-kappa-B p105 subunit, Nuclear factor of kappa light polypeptide gene enhancer in B-cells 1, p105 | |
Goat | |
Affinity Purified | |
RUO | |
4790 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
ChIP Assay | |
LYOPH | |
Western Blot 0.5 ug/mL, Chromatin Immunoprecipitation (ChIP) 5 ug/5 x 10^6 cells | |
NP_003989 | |
NFKB1 | |
E. coli-derived recombinant human NF kappa B1 Pro317-Asp437 Accession # NP_003989 | |
25 μg | |
Primary | |
Detects endogenous human and mouse NF kappa B1 in Western blots. | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction