Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse NGFRAP1/BEX3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB6858
This item is not returnable.
View return policy
Description
NGFRAP1/BEX3/NADE Monoclonal specifically detects NGFRAP1/BEX3/NADE in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
NGFRAP1/BEX3/NADE | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL | |
Q00994 | |
NGFRAP1 | |
E. coli-derived recombinant human NGFRAP1/BEX3 Met1-Pro111 Accession # Q00994 | |
100 μg | |
Primary | |
Detects human NGFRAP1/BEX3 in direct ELISAs. | |
Human, Mouse | |
Purified |
Western Blot, Immunohistochemistry | |
672911 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
BEX3, BEX3p75NTR-associated cell death executor, Brain-expressed X-linked protein 3, DXS6984EX-linked 3, HGR74, NADE, nerve growth factor receptor (TNFRSF16) associated protein 1, Nerve growth factor receptor-associated protein 1, Ovarian granulosa cell 13.0 kDa protein HGR74, ovarian granulosa cell protein (13kD), protein BEX3 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
27018 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction