Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse Nogo-B Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF6034
missing translation for 'nonReturnableMessage'
missing translation for 'returnPolicyLabel'
Description
Nogo-B Polyclonal specifically detects Nogo-B in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Nogo-B | |
Polyclonal | |
Unconjugated | |
NP_722550 | |
Sheep | |
Antigen Affinity-purified | |
RUO | |
57142 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
RTN4 | |
Chinese hamster ovary cell line CHO-derived recombinant human Nogo-B Met1-Val200 Accession # NP_722550 | |
100 μg | |
Primary | |
Detects human and mouse Nogo-B in direct ELISAs and Western blots. | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction