Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse Nurr1/NGFI-B beta/NR4A2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF2156
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF2156 100 μg
AF2156SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF2156 Supplier R&D Systems Supplier No. AF2156
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 5 publications

Nurr1/NGFI-B beta/NR4A2 Polyclonal specifically detects Nurr1/NGFI-B beta/NR4A2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Nurr1/NGFI-B beta/NR4A2
Applications Western Blot, Immunohistochemistry, Immunocytochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunocytochemistry 5-15 ug/mL
Gene Accession No. Q06219
Gene Alias human homolog of, NGFI-B beta, NOT, NR4A2, nuclear receptor subfamily 4, group A, member 2, orphan nuclear receptor NR4A2, Orphan nuclear receptor NURR1, T-cell nuclear receptor NOT, TINUR, transcriptionally inducible nuclear receptor related 1, Transcriptionally-inducible nuclear receptor
Gene Symbols NR4A2
Host Species Goat
Immunogen E. coli-derived recombinant mouse Nurr1/NGFI-B beta/NR4A2 Val332-Lys558 Accession # Q06219
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4929
Test Specificity Detects human and mouse Nurr1/NGFI-B beta/NR4A2 in direct ELISAs and Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.