Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse Oct-3/4 Alexa Fluor 405-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems IC1759V100UG
This item is not returnable.
View return policy
Description
Oct-3/4 Monoclonal specifically detects Oct-3/4 in Human, Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
Oct-3/4 | |
Monoclonal | |
0.2 mg/mL | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
Q01860 | |
Rat | |
100 μg | |
Primary | |
Detects human Oct-3/4 in Western blots and detects mouse Oct-3/4 in flow cytometry. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Flow Cytometry | |
240408 | |
Alexa Fluor 405 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
POU5F1 | |
E. coli-derived recombinant human Oct-3/4 Met1-Asn265 (Met262Leu) Accession # Q01860 | |
RUO | |
5460 | |
Human, Mouse | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction