Learn More
Human/Mouse Oct-3/4 Antibody, R&D Systems™
GREENER_CHOICE
Goat Polyclonal Antibody has been used in 29 publications
Supplier:  R&D Systems AF1759
Catalog No. AF1759
Only null left
Add to Cart
This item is not returnable. View return policy
Description
Description
Oct-3/4 Polyclonal specifically detects Oct-3/4 in Human, Mouse samples. It is validated for Western Blot, Simple Western, Immunoprecipitation, Chromatin Immunoprecipitation (ChIP), Immunocytochemistry.
Specifications
Specifications
Oct-3/4
Polyclonal
Unconjugated
Q01860
Goat
Affinity Purified
RUO
5460
Reconstitute at 0.2 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
ChIP Assay, Immunocytochemistry, Immunoprecipitation
LYOPH
Western Blot 0.5 ug/mL, Simple Western 20 ug/mL, Immunoprecipitation 3 ug/10^6 cells, Chromatin Immunoprecipitation (ChIP) 5 ug/10^6 cells, Immunocytochemistry 5-15 ug/mL
POU5F1
E. coli-derived recombinant human Oct-3/4 Met1-Asn265 (Met262Leu) Accession # Q01860
100 μg
Primary
Detects human Oct-3/4 in direct ELISAs and Western blots.
Human, Mouse
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.