Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse Oct-3/4 PE-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 9 publications
$221.50 - $444.00
Specifications
Antigen | Oct-3/4 |
---|---|
Clone | 240408 |
Dilution | Intracellular Staining by Flow Cytometry 10 uL/10^6 cells |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
IC1759P025
![]() |
R&D Systems
IC1759P025 |
25 Tests |
Each for $221.50
|
|
|||||
IC1759P100
![]() |
R&D Systems
IC1759P100 |
100 Tests |
Each for $444.00
|
|
|||||
Description
Oct-3/4 Monoclonal specifically detects Oct-3/4 in Human, Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
Oct-3/4 | |
Intracellular Staining by Flow Cytometry 10 uL/10^6 cells | |
Monoclonal | |
Purified | |
RUO | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
5460 | |
E. coli-derived recombinant human Oct-3/4 Met1-Asn265 (Met262Leu) Accession # Q01860 | |
Primary | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
240408 | |
Flow Cytometry | |
R-PE | |
Rat | |
Human, Mouse | |
Q01860 | |
POU5F1 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Detects human Oct-3/4 in Western blots and detects mouse Oct-3/4 in flow cytometry. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title