Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse OMgp Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB1674SP
Change view
Click to view available options
Quantity:
25 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1674SP 25 μg
MAB1674 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB1674SP Supplier R&D Systems Supplier No. MAB1674SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

OMgp Monoclonal specifically detects OMgp in Human, Mouse samples. It is validated for Western Blot.

Specifications

Antigen OMgp
Applications Western Blot
Classification Monoclonal
Clone 240527
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q63912
Gene Alias oligodendrocyte myelin glycoprotein, OMG, OMGPoligodendrocyte-myelin glycoprotein
Gene Symbols OMG
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse OMgp Ile25-Ser420 Accession # Q63912
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4974
Test Specificity Detects human and mouse OMgp in direct ELISAs and Western blots. In direct ELISAs and Western blots, 100% cross-reactivity with recombinant human OMgp is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.