Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse Orexin A/Hypocretin-1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB763100
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB763100 100 μg
MAB763SP 25 μg
MAB763500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB763100 Supplier R&D Systems Supplier No. MAB763100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Orexin A/Hypocretin-1 Monoclonal antibody specifically detects Orexin A/Hypocretin-1 in Human, Mouse samples. It is validated for Immunohistochemistry.

Specifications

Antigen Orexin A/Hypocretin-1
Applications Immunohistochemistry
Classification Monoclonal
Clone 97505
Conjugate Unconjugated
Dilution Immunohistochemistry 5-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. O43612
Gene Alias HCRT, Hcrt1, hypocretin (orexin) neuropeptide precursor, Hypocretin1, Hypocretin-1, NRCLP1, orexin, OXHcrt, PPORX, PPOXHypocretin
Gene Symbols HCRT
Host Species Mouse
Immunogen KLH-coupled human Orexin A/Hypocretin-1 synthetic peptide Accession # O43612
Purification Method Protein A or G purified from ascites
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3060
Test Specificity Detects human and mouse Orexin A/Hypocretin-1. In dot blots, no cross-reactivity with recombinant human Orexin B is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.