Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse Osteocrin Antibody, R&D Systems™

Rat Monoclonal Antibody
$200.50 - $502.50
Specifications
Antigen | Osteocrin |
---|---|
Clone | 311413 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB26201SP
![]() |
R&D Systems
MAB26201SP |
25 μg |
Each for $200.50
|
|
|||||
MAB26201
![]() |
R&D Systems
MAB26201 |
100 μg |
Each for $502.50
|
|
|||||
Description
Osteocrin Monoclonal specifically detects Osteocrin in Human, Mouse samples. It is validated for Western Blot.Specifications
Osteocrin | |
LYOPH | |
Western Blot | |
Unconjugated | |
Rat | |
Human, Mouse | |
P61364 | |
344901 | |
E. coli-derived recombinant mouse Osteocrin Val28-Gly130 Accession # P61364 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects only the pro form (11 kDa) of human and mouse Osteocrin in direct ELISAs and Western blots. |
311413 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
MUSCLIN, osteocrin, OSTN | |
OSTN | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title