Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse P-Cadherin APC-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 2 publications
Supplier: R&D Systems FAB761A
This item is not returnable.
View return policy
Description
P-Cadherin Monoclonal specifically detects P-Cadherin in Human, Mouse samples. It is validated for Flow Cytometry.Specifications
P-Cadherin | |
Monoclonal | |
APC | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
CAD3, cadherin 3, P-cadherin (placental), cadherin 3, type 1, P-cadherin (placental), cadherin-3, CDH3, CDHP, CDHPcalcium-dependent adhesion protein, placental, HJMD, PCAD, PCadherin, PCADP-cadherin, Placental cadherin | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
1001 | |
Human, Mouse | |
Purified |
Flow Cytometry | |
106020 | |
Flow Cytometry 10 uL/10^6 cells | |
P10287 | |
CDH3 | |
Mouse myeloma cell line NS0-derived recombinant mouse P-Cadherin Glu100-Gly647 Accession # P10287 | |
100 Tests | |
Primary | |
Detects mouse P-Cadherin in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) N-Cadherin, recombinant mouse (rm) VE-Cadherin, rhCadherin-8, or rhCadherin-17 is observed. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction