Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse PAK3 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF6897
This item is not returnable.
View return policy
Description
PAK3 Polyclonal specifically detects PAK3 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
PAK3 | |
Polyclonal | |
Unconjugated | |
O75914 | |
PAK3 | |
E. coli-derived recombinant human PAK3 Leu138-Gln255 Accession # O75914 | |
100 μg | |
Primary | |
Detects human PAK3 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) PAK1, rhPAK1B, rhPAK2, rhPAK4, rhPAK6, and rhPAK7 is observed. | |
Human, Mouse | |
IgG |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 0.25 ug/mL, Immunohistochemistry 5-15 ug/mL | |
beta-PAK, bPAK, CDKN1A, EC 2.7.11, EC 2.7.11.1, hPAK3, mental retardation, X-linked 47, MRX30, MRX47, Oligophrenin-3, OPHN3, p21 (CDKN1A)-activated kinase 3, p21 protein (Cdc42/Rac)-activated kinase 3, p21-activated kinase 3, PAK-3, PAK3beta, serine/threonine-protein kinase PAK 3 | |
Sheep | |
Affinity Purified | |
RUO | |
5063 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction