Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse Park7/DJ-1 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF3668
This item is not returnable.
View return policy
Description
Park7/DJ-1 Polyclonal specifically detects Park7/DJ-1 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry, Knockout Validated.Specifications
Park7/DJ-1 | |
Polyclonal | |
Unconjugated | |
Q99LX0 | |
PARK7 | |
E. coli-derived recombinant mouse Park7/DJ-1 Ala2-Asp189 Accession # Q99LX0 | |
100 μg | |
Primary | |
Detects human and mouse Park7/DJ-1 in direct ELISAs and Western blots. | |
Human, Mouse | |
IgG |
Western Blot, Immunocytochemistry, KnockDown | |
LYOPH | |
Western Blot 0.5 ug/mL, Immunocytochemistry 5-15 ug/mL, Knockout Validated | |
DJ1, DJ-1, DJ1FLJ34360, EC 3.4, FLJ27376, FLJ92274, Oncogene DJ1, Parkinson disease (autosomal recessive, early onset) 7, Parkinson disease protein 7, protein DJ-1 | |
Goat | |
Affinity Purified | |
RUO | |
11315 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction