Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse PARP Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 5 publications
Supplier: R&D Systems AF600SP
This item is not returnable.
View return policy
Description
PARP Polyclonal specifically detects PARP in Human, Mouse samples. It is validated for Western Blot, Simple Western, Immunoprecipitation, Immunocytochemistry, Knockout Validated.Specifications
PARP | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
ADP-ribosyltransferase (NAD+; poly (ADP-ribose) polymerase), ADPRT, ADPRT 1, ADPRTADP-ribosyltransferase NAD(+), EC 2.4.2, EC 2.4.2.30, NAD(+) ADP-ribosyltransferase 1, PARP apoptosis, PARP1, PARP-1, PARPADPRT1, poly (ADP-ribose) polymerase 1, poly (ADP-ribose) polymerase family, member 1, poly [ADP-ribose] polymerase 1, poly(ADP-ribose) polymerase, poly(ADP-ribose) synthetase, poly(ADP-ribosyl)transferase, Poly[ADP-ribose] synthase 1, PPOL, PPOLpADPRT-1 | |
Goat | |
Affinity Purified | |
RUO | |
142 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunoprecipitation, Immunocytochemistry, KnockDown | |
LYOPH | |
Western Blot 0.4 ug/mL, Simple Western 5 ug/mL, Immunoprecipitation 5 ug/10^6 cells, Immunocytochemistry 1-25 ug/mL, Knockout Validated | |
NP_031441 | |
PARP1 | |
E. coli-derived recombinant mouse PARP Val71-Pro329 Accession # NP_031441 | |
25 μg | |
Primary | |
Detects human and mouse PARP in Western blots. | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction