Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse Pax5/BSAP Alexa Fluor 350-conjugated Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Supplier: R&D Systems IC3487U100UG
This item is not returnable.
View return policy
Description
Pax5/BSAP Monoclonal specifically detects Pax5/BSAP in Human, Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
Pax5/BSAP | |
Monoclonal | |
Alexa Fluor 350 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
B cell specific activator protein, B-cell-specific transcription factor, BSAP, BSAPB-cell lineage specific activator, paired box 5, paired box gene 5 (B-cell lineage specific activator protein), paired box gene 5 (B-cell lineage specific activator), paired box homeotic gene 5, paired box protein Pax-5, transcription factor PAX 5 | |
E. coli-derived recombinant human Pax5 Thr141-His391 Accession # Q02548 | |
RUO | |
5079 | |
Human, Mouse | |
IgG |
Flow Cytometry | |
1207C | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
Q02548 | |
Rabbit | |
100 μg | |
Primary | |
Detects human Pax5 in direct ELISAs and Western blots. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction