Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse PBR Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$214.00 - $491.00
Specifications
Antigen | PBR |
---|---|
Clone | 2393B |
Dilution | Western Blot 1 μg/mL, Immunohistochemistry 3-25 μg/mL, Immunocytochemistry 3-25 μg/mL |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB10045SP
![]() |
R&D Systems
MAB10045SP |
25 μg |
Each for $214.00
|
|
|||||
MAB10045100
![]() |
R&D Systems
MAB10045100 |
100 μg |
Each for $491.00
|
|
|||||
Description
PBR Monoclonal specifically detects PBR in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry.Specifications
PBR | |
Western Blot 1 μg/mL, Immunohistochemistry 3-25 μg/mL, Immunocytochemistry 3-25 μg/mL | |
Monoclonal | |
Supernatant | |
Human, Mouse | |
P30536 | |
706.0 | |
IgG | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
2393B | |
Western Blot, Immunohistochemistry, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
benzodiazepine peripheral binding site, BZRPDBI, IBP, MBRPBS, mDRC, Mitochondrial benzodiazepine receptor, PBRBPBS, PBRS, peripheral benzodiazepine receptor, Peripheral-type benzodiazepine receptor, pk18, PKBSbenzodiazapine receptor (peripheral), PTBR, translocator protein, translocator protein (18kDa), TSPO | |
Synthetic peptide containing human PBR, Accession No. P30536 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human PBR in direct ELISAs. Detects human and mouse PBR in Western blots. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title