Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse Periostin/OSF-2 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 4 publications
Supplier: R&D Systems MAB3548SP
This item is not returnable.
View return policy
Description
Periostin/OSF-2 Monoclonal specifically detects Periostin/OSF-2 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry.Specifications
Periostin/OSF-2 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL | |
NP_056599 | |
POSTN | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse Periostin isoform 2 Asn24-Gln811 Accession # NP_056599 | |
25 μg | |
Primary | |
Detects mouse Periostin/OSF-2 in direct ELISAs and Western blots. | |
Human, Mouse | |
Purified |
Western Blot, Immunocytochemistry | |
345613 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Fasciclin I-like, MGC119510, MGC119511, OSF2, OSF-2, OSF-2osteoblast specific factor 2 (fasciclin I-like), OSF2periodontal ligament-specific periostin, Osteoblast-specific factor 2, PDLPOSTN, periostin, periostin isoform thy2, periostin isoform thy4, periostin isoform thy6, periostin isoform thy8, periostin, osteoblast specific factor, PNRP11-412K4.1, POSTN, TRIF52 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
10631 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction