Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse Peroxiredoxin 3 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Supplier: R&D Systems AF6610SP
This item is not returnable.
View return policy
Description
Peroxiredoxin 3 Polyclonal specifically detects Peroxiredoxin 3 in Human, Mouse samples. It is validated for Western Blot.Specifications
| Peroxiredoxin 3 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Antioxidant protein 1MGC104387, AOP-1, AOP-1MGC24293, AOP1PRO1748, EC 1.11.1.15, HBC189, MER5, peroxiredoxin 3, Peroxiredoxin III, peroxiredoxin-3, PRDX3, Protein MER5 homolog, prx-III, SP-22, thioredoxin-dependent peroxide reductase, mitochondrial | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 10935 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| P20108 | |
| PRDX3 | |
| E. coli-derived recombinant mouse Peroxiredoxin 3 Met1-Gln257 Accession # P20108 | |
| 25 μg | |
| Primary | |
| Detects human and mouse Peroxiredoxin 3 in direct ELISAs and Western blots. In direct ELISAs, approximately 6% cross-reactivity with recombinant mouse (rm) Peroxiredoxin 2 is observed and less than 1% cross-reactivity with recombinant human (rh) Peroxiredoxin 1, rmPeroxiredoxin 5, and rmPeroxiredoxin 6 is observed. | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction