Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse Peroxiredoxin 3 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF6610
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF6610 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF6610 Supplier R&D Systems Supplier No. AF6610
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 2 publications

Peroxiredoxin 3 Polyclonal specifically detects Peroxiredoxin 3 in Human, Mouse samples. It is validated for Western Blot.

Specifications

Antigen Peroxiredoxin 3
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Gene Accession No. P20108
Gene Alias Antioxidant protein 1MGC104387, AOP-1, AOP-1MGC24293, AOP1PRO1748, EC 1.11.1.15, HBC189, MER5, peroxiredoxin 3, Peroxiredoxin III, peroxiredoxin-3, PRDX3, Protein MER5 homolog, prx-III, SP-22, thioredoxin-dependent peroxide reductase, mitochondrial
Gene Symbols PRDX3
Host Species Goat
Immunogen E. coli-derived recombinant mouse Peroxiredoxin 3 Met1-Gln257 Accession # P20108
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10935
Test Specificity Detects human and mouse Peroxiredoxin 3 in direct ELISAs and Western blots. In direct ELISAs, approximately 6% cross-reactivity with recombinant mouse (rm) Peroxiredoxin 2 is observed and less than 1% cross-reactivity with recombinant human (rh) Peroxiredoxin 1, rmPeroxiredoxin 5, and rmPeroxiredoxin 6 is observed.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Human, Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.