Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse PGPEP-1 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF6278
This item is not returnable.
View return policy
Description
PGPEP-1 Polyclonal specifically detects PGPEP-1 in Human, Mouse samples. It is validated for Western Blot.Specifications
PGPEP-1 | |
Polyclonal | |
Unconjugated | |
Q9NXJ5 | |
PGPEP1 | |
E. coli-derived recombinant human PGPEP-1 Met1-His209 Accession # Q9NXJ5 | |
100 μg | |
Primary | |
Detects human PGPEP-1 in direct ELISAs and, human and mouse PGPEP-1 in Western blots. | |
Human, Mouse | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
EC 3.4.19.3, PAP-I, PCP, PGPEP1, PGPIFLJ20208, PGP-IPAP-I, PGPMGC10812, Pyroglutamyl aminopeptidase I, pyroglutamyl-peptidase 1, pyroglutamyl-peptidase I5-oxoprolyl-peptidase, Pyrrolidone-carboxylate peptidase | |
Sheep | |
Affinity Purified | |
RUO | |
54858 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction