Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse Phospho-ERK1 (ERK1 T202/Y204, ERK2 T185/Y187) Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Supplier: R&D Systems MAB18251SP
This item is not returnable.
View return policy
Description
ERK1 Monoclonal specifically detects ERK1 in Human, Mouse samples. It is validated for Western Blot, Simple Western.Specifications
ERK1 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide | |
EC 2.7.11, ERK-1, ERK1p44-MAPK, ERT2, Extracellular signal-regulated kinase 1, extracellular signal-related kinase 1, HS44KDAP, HUMKER1A, Insulin-stimulated MAP2 kinase, MAP kinase 1, MAP kinase 3, MAP kinase isoform p44, MAPK 1, MAPK 3, MAPK3, MGC20180, Microtubule-associated protein 2 kinase, Mitogen-activated protein kinase 1, mitogen-activated protein kinase 3, p44erk1, p44-ERK1, P44MAPK, PRKM3, PRKM3EC 2.7.11.24 | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
RUO | |
5595 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG |
Western Blot | |
1088B | |
Western Blot 0.1 ug/mL, Simple Western 1 ug/mL | |
P27361 | |
MAPK3 | |
Phosphopeptide containing the human ERK1 T202/Y204 site Accession # P27361 | |
25 μg | |
Primary | |
Detects human and mouse ERK1 and ERK2 dually phosphorylated at T202/Y204 or T185/Y187, respectively. | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction