Learn More
Human/Mouse Phospho-HGFR/c-MET (Y1349) Antibody, R&D Systems™
GREENER_CHOICE
Rabbit Polyclonal Antibody
Supplier:  R&D Systems AF3950
Catalog No. AF3950
Only null left
Add to Cart
This item is not returnable. View return policy
Description
Description
HGFR/c-MET Polyclonal specifically detects HGFR/c-MET in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry.
Specifications
Specifications
HGFR/c-MET (p tyr1349)
Polyclonal
Unconjugated
AUTS9, c-Met, cMET, EC 2.7.10, EC 2.7.10.1, hepatocyte growth factor receptor, HGF R, HGF receptor, HGF/SF receptor, HGFR, MET, Met (c-Met), met proto-oncogene (hepatocyte growth factor receptor), met proto-oncogene tyrosine kinase, oncogene MET, Proto-oncogene c-Met, RCCP2, Scatter factor receptor, SF receptor, Tyrosine-protein kinase Met
Rabbit
Affinity Purified
RUO
4233
Reconstitute at 0.2 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Western Blot, Immunohistochemistry, Immunocytochemistry
LYOPH
Western Blot 0.5 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunocytochemistry 5-15 ug/mL
MET
Phosphopeptide containing human HGF R Y1349 site
50 μg
Primary
Detects human and mouse HGF R/c-MET when phosphorylated at Y1349.
Human, Mouse
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.