Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse Phospho- Histone H2AX (S139) Antibody, R&D Systems™

Catalog No. 4418APC020
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-18A-PC020 20 μL
44-18A-PC100 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 44-18A-PC020 Supplier R&D Systems Supplier No. 4418APC020
This item is not returnable. View return policy

Rabbit Antibody

Histone H2AX specifically detects Histone H2AX in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry.

Specifications

Antigen Histone H2AX
Applications Western Blot, Immunocytochemistry
Conjugate Unconjugated
Dilution Western Blot 1:2500 dilution, Immunocytochemistry 1.7-15 ug/mL
Formulation Supplied as a solution containing PBS.
Gene Alias H2A.X, H2A/X, H2AFX
Host Species Rabbit
Immunogen Phosphopeptide containing human Histone H2AX S139 site
Purification Method Antigen Affinity-purified
Quantity 20 μL
Primary or Secondary Primary
Gene ID (Entrez) 3014
Test Specificity Detects human and mouse Histone H2AX when phosphorylated at S139 in Western blots. This antibody detects only double-strand DNA breaks.
Target Species Human, Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C, as supplied. 1 month, 2 to 8 °C under sterile conditions after opening. 6 months, -20 to -70 °C under sterile conditions after opening.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.