Learn More
Human/Mouse Phospho- Histone H2AX (S139) Antibody, R&D Systems™
Description
Specifications
Specifications
| Antigen | Histone H2AX |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:2500 dilution, Immunocytochemistry 1.7-15 ug/mL |
| Formulation | Supplied as a solution containing PBS. |
| Gene Alias | H2A.X, H2A/X, H2AFX |
| Immunogen | Phosphopeptide containing human Histone H2AX S139 site |
| Purification Method | Antigen Affinity-purified |
| Quantity | 100 μL |
| Primary or Secondary | Primary |
| Gene ID (Entrez) | 3014 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.