Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse Phospho-PDGF R beta (Y751) Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Supplier: R&D Systems MAB9027
This item is not returnable.
View return policy
Description
PDGF R beta Monoclonal specifically detects PDGF R beta in Human, Mouse samples. It is validated for Western Blot, Simple Western, Immunocytochemistry.Specifications
PDGF R beta | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide | |
beta-type platelet-derived growth factor receptor, CD140 antigen-like family member B, CD140b, CD140b antigen, EC 2.7.10, EC 2.7.10.1, JTK12, PDGFR, PDGFR1, PDGFRB, PDGF-R-beta, platelet-derived growth factor receptor, beta polypeptide | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
RUO | |
5159 | |
Reconstitute at 0.5 mg/mL in sterile PBS | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG |
Western Blot, Immunocytochemistry | |
1210B | |
Western Blot 0.1 ug/mL, Simple Western 1 ug/mL, Immunocytochemistry 5-25 ug/mL | |
P09619 | |
PDGFRB | |
Phosphopeptide containing the human PDGF R beta (Y751) site Accession # P09619 | |
100 μg | |
Primary | |
Detects human and mouse PDGF R beta when phosphorylated at Y751 in Western blots. | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction