Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse Phospho-RSK1 (S221)/RSK2 (S227) Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB892
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB892 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB892 Supplier R&D Systems Supplier No. MAB892
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

RSK1/RSK2 Monoclonal specifically detects RSK1/RSK2 in Human, Mouse samples. It is validated for Western Blot, Simple Western.

Specifications

Antigen RSK1/RSK2
Applications Western Blot
Classification Monoclonal
Clone 1019B
Conjugate Unconjugated
Dilution Western Blot 1:1000 dilution, Simple Western 1:100 dilution
Formulation Supplied as a solution in PBS containing BSA, Glycerol and Sodium Azide. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide
Host Species Rabbit
Immunogen Phosphopeptide containing the human RSK1 S221 site
Purification Method Protein A or G purified from cell culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Detects human and mouse RSK1 when phosphorylated at S221, and human and mouse RSK2 when phosphorylated at S227.
Target Species Human, Mouse
Content And Storage 12 months from date of receipt, -20 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after opening. 6 months, -20 degreesC under sterile conditions after opening.
Form Purified
Isotype IgG
Show More Show Less

Contains <0.1% Sodium Azide, which is not hazardous at this concentration according to GHS classifications. Refer to SDS for additional information and handling instructions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.