Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse Pin1 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 6 publications
Supplier: R&D Systems MAB2294
This item is not returnable.
View return policy
Description
Pin1 Monoclonal specifically detects Pin1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry.Specifications
Pin1 | |
Monoclonal | |
LYOPH | |
Western Blot 0.5 ug/mL, Immunohistochemistry 8-25 ug/mL, Immunoprecipitation 2-3 ug/500 ug cell lysate, Immunocytochemistry 8-25 ug/mL | |
Q13526 | |
PIN1 | |
E. coli-derived recombinant human Pin1 Ala2-Glu163 Accession # Q13526 | |
100 μg | |
Primary | |
Detects human and mouse Pin1 in Western blots. | |
Human, Mouse | |
Purified |
Western Blot, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry | |
257417 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
dod, EC 5.2.1.8, peptidyl-prolyl cis/trans isomerase, NIMA-interacting, peptidylprolyl cis/trans isomerase, NIMA-interacting 1, peptidyl-prolyl cis-trans isomerase NIMA-interacting 1, Peptidyl-prolyl cis-trans isomerase Pin1, PPIase Pin1, prolyl isomerase, protein (peptidyl-prolyl cis/trans isomerase) NIMA-interacting 1, protein (peptidylprolyl cis/trans isomerase) NIMA-interacting 1, Rotamase Pin1, UBL5 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
5300 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction