Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse PKA RII alpha Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB8000100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB8000100 100 μg
MAB8000SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB8000100 Supplier R&D Systems Supplier No. MAB8000100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Protein Kinase A Regulatory Subunit II alpha Monoclonal specifically detects Protein Kinase A Regulatory Subunit II alpha in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry.

Specifications

Antigen Protein Kinase A Regulatory Subunit II alpha
Applications Western Blot, Immunohistochemistry, Immunocytochemistry
Classification Monoclonal
Clone 2394D
Conjugate Unconjugated
Dilution Western Blot 0.5 μg/mL, Immunohistochemistry 0.3-25 μg/mL, Immunocytochemistry 8-25 μg/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. P13861
Host Species Rabbit
Immunogen Synthetic peptide containing Human/Mouse PKA RII alpha , Accession # P13861
Purification Method Protein A or G purified from cell culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Test Specificity Detects human PKA RII alpha in direct ELISAs. Detects human and mouse PKA RII alpha in Western blots.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.