Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse PKC alpha Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB5340
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5340 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB5340 Supplier R&D Systems Supplier No. MAB5340
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

PKC alpha Monoclonal specifically detects PKC alpha in Human, Mouse samples. It is validated for Western Blot.

Specifications

Antigen PKC alpha
Applications Western Blot
Classification Monoclonal
Clone 421810
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P17252
Gene Alias AAG6, aging-associated gene 6, EC 2.7.11, EC 2.7.11.13, MGC129901, PKCA, PKC-A, PKC-alpha, PKCAMGC129900, PRKACA, PRKCA, protein kinase C alpha type, protein kinase C, alpha
Gene Symbols PRKCA
Host Species Rat
Immunogen E. coli-derived recombinant human PKC alpha Lys604-Val672 Accession # P17252
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5578
Test Specificity Detects endogenous human and mouse PKC alpha in Western blots.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.