Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse PKN1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB6100SP
This item is not returnable.
View return policy
Description
PKN1 Monoclonal specifically detects PKN1 in Human, Mouse samples. It is validated for Western Blot.Specifications
PKN1 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL | |
Q16512 | |
PKN1 | |
E. coli-derived recombinant human PKN1 Ile215-Thr388 Accession # Q16512 | |
25 μg | |
Primary | |
Detects human and mouse PKN1 in Western blots. | |
Human, Mouse | |
Purified |
Western Blot | |
540421 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
DBK, EC 2.7.11, MGC46204, PAK1, PAK1Serine-threonine protein kinase N, PKN, PKN-ALPHA, PKNPKN-ALPHA, PRK1, PRK1PAK-1, PRKCL1, PRKCL1protein kinase C-related kinase 1, Protease-activated kinase 1, Protein kinase C-like 1EC 2.7.11.13, Protein kinase C-like PKN, protein kinase N1, Protein kinase PKN-alpha, Protein-kinase C-related kinase 1, serine/threonine protein kinase N, serine/threonine-protein kinase N1, serine-threonine kinase N | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
5585 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction